Merck
  • Aberrant DNA methylation of WNT pathway genes in the development and progression of CIMP-negative colorectal cancer.
Aberrant DNA methylation of WNT pathway genes in the development and progression of CIMP-negative colorectal cancer.
Epigenetics (2016-06-02)
Orsolya Galamb, Alexandra Kalmár, Bálint Péterfia, István Csabai, András Bodor, Dezső Ribli, Tibor Krenács, Árpád V Patai, Barnabás Wichmann, Barbara Kinga Barták, Kinga Tóth, Gábor Valcz, Sándor Spisák, Zsolt Tulassay, Béla Molnár
ABSTRACT
The WNT signaling pathway has an essential role in colorectal carcinogenesis and progression, which involves a cascade of genetic and epigenetic changes. We aimed to analyze DNA methylation affecting the WNT pathway genes in colorectal carcinogenesis in promoter and gene body regions using whole methylome analysis in 9 colorectal cancer, 15 adenoma, and 6 normal tumor adjacent tissue (NAT) samples by methyl capture sequencing. Functional methylation was confirmed on 5-aza-2'-deoxycytidine-treated colorectal cancer cell line datasets. In parallel with the DNA methylation analysis, mutations of WNT pathway genes (APC, β-catenin/CTNNB1) were analyzed by 454 sequencing on GS Junior platform. Most differentially methylated CpG sites were localized in gene body regions (95% of WNT pathway genes). In the promoter regions, 33 of the 160 analyzed WNT pathway genes were differentially methylated in colorectal cancer vs. normal, including hypermethylated AXIN2, CHP1, PRICKLE1, SFRP1, SFRP2, SOX17, and hypomethylated CACYBP, CTNNB1, MYC; 44 genes in adenoma vs. NAT; and 41 genes in colorectal cancer vs. adenoma comparisons. Hypermethylation of AXIN2, DKK1, VANGL1, and WNT5A gene promoters was higher, while those of SOX17, PRICKLE1, DAAM2, and MYC was lower in colon carcinoma compared to adenoma. Inverse correlation between expression and methylation was confirmed in 23 genes, including APC, CHP1, PRICKLE1, PSEN1, and SFRP1. Differential methylation affected both canonical and noncanonical WNT pathway genes in colorectal normal-adenoma-carcinoma sequence. Aberrant DNA methylation appears already in adenomas as an early event of colorectal carcinogenesis.
Product Description
Roche
High Pure PCR Template Preparation Kit, pkg of 100 purifications, suitable for DNA extraction