Merck
  • Identification of ellagic acid as potent inhibitor of protein kinase CK2: a successful example of a virtual screening application.
Identification of ellagic acid as potent inhibitor of protein kinase CK2: a successful example of a virtual screening application.
Journal of medicinal chemistry (2006-04-14)
Giorgio Cozza, Paolo Bonvini, Elisa Zorzi, Giorgia Poletto, Mario A Pagano, Stefania Sarno, Arianna Donella-Deana, Giuseppe Zagotto, Angelo Rosolen, Lorenzo A Pinna, Flavio Meggio, Stefano Moro
ABSTRACT
Casein kinase 2 (CK2) is a ubiquitous, essential, and highly pleiotropic protein kinase whose abnormally high constitutive activity is suspected to underlie its pathogenic potential in neoplasia and other diseases. Using a virtual screening approach, we have identified the ellagic acid, a naturally occurring tannic acid derivative, as a novel potent CK2 inhibitor. At present, ellagic acid represents the most potent known CK2 inhibitor (K(i) = 20 nM).
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark