Merck
  • Protein phosphatase 1 catalyzes HBV core protein dephosphorylation and is co-packaged with viral pregenomic RNA into nucleocapsids.
Protein phosphatase 1 catalyzes HBV core protein dephosphorylation and is co-packaged with viral pregenomic RNA into nucleocapsids.
PLoS pathogens (2020-07-24)
Zhanying Hu, Haiqun Ban, Haiyan Zheng, Mingliang Liu, Jinhong Chang, Ju-Tao Guo
ABSTRACT
Hepatitis B virus (HBV) replicates its genomic DNA via viral DNA polymerase self-primed reverse transcription of a RNA pre-genome in the nucleocapsid assembled by 120 core protein (Cp) dimers. The arginine-rich carboxyl-terminal domain (CTD) of Cp plays an important role in the selective packaging of viral DNA polymerase-pregenomic (pg) RNA complex into nucleocapsid. Previous studies suggested that the CTD is initially phosphorylated at multiple sites to facilitate viral RNA packaging and subsequently dephosphorylated in association with viral DNA synthesis and secretion of DNA-containing virions. However, our recent studies suggested that Cp is hyper-phosphorylated as free dimers and its dephosphorylation is associated with pgRNA encapsidation. Herein, we provide further genetic and biochemical evidence supporting that extensive Cp dephosphorylation does take place during the assembly of pgRNA-containing nucleocapsids, but not empty capsids. Moreover, we found that cellular protein phosphatase 1 (PP1) is required for Cp dephosphorylation and pgRNA packaging. Interestingly, the PP1 catalytic subunits α and β were packaged into pgRNA-containing nucleocapsids, but not empty capsids, and treatment of HBV replicating cells with core protein allosteric modulators (CpAMs) promoted empty capsid assembly and abrogated the encapsidation of PP1 α and β. Our study thus identified PP1 as a host cellular factor that is co-packaged into HBV nucleocapsids, and plays an essential role in selective packaging of the viral DNA-polymerase-pgRNA complex through catalyzing Cp dephosphorylation.
Sigma-Aldrich
PP1, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)