Merck
  • VIP Regulates Morphology and F-Actin Distribution of Schlemm's Canal in a Chronic Intraocular Pressure Hypertension Model via the VPAC2 Receptor.
VIP Regulates Morphology and F-Actin Distribution of Schlemm's Canal in a Chronic Intraocular Pressure Hypertension Model via the VPAC2 Receptor.
Investigative ophthalmology & visual science (2018-07-20)
Liwen Chen, Mu Li, Zhaoxia Luo, Xiaoqin Yan, Ke Yao, Yin Zhao, Hong Zhang
ABSTRACT
To investigate the roles of vasoactive intestinal peptides (VIPs) in regulating the morphology and F-actin distribution of Schlemm's canal (SC) of rat eyes. Chronic intraocular pressure (IOP) hypertension models with episcleral venous cauterization (EVC) were treated with topical VIP or PG99-465 (vasoactive intestinal peptide receptors 2 [VPAC2] antagonist). IOPs were measured with Tono-Pen, and the SC parameters, including the cross-section area, circumference, and length, were statistically evaluated by hematoxylin-eosin and CD31 immunohistochemical staining. Immunofluorescence was performed to detect the distribution of F-actin in the SC. Moreover, the distribution of filamentous actin (F-actin) and globular actin (G-actin) in human umbilical vein endothelial cells (HUVECs) was studied under a pressure system by immunofluorescence and Western blotting. Increased expressions of VIP and VPAC2 receptors, as well as a disordered distribution of F-actin were found in SC endothelial cells (SCEs) in the EVC model. Moreover, topical VIP maintained the normal distribution of F-actin in SCEs, expanded the collapsed SC, and induced a significant decrease in IOP in the EVC model. In in vitro HUVECs, the F-actin/G-actin ratio increased significantly under stress stimulation for 30 minutes. A total of 50 μM VIP helped maintain the normal F-actin/G-actin ratio of HUVECs against stress stimulation. VIP regulates the distribution of F-actin in SCEs via the VPAC2 receptor in order to induce a decrease in IOP. VIP may represent a new target for antiglaucoma drugs.