Merck
  • SnS2 /Co3 S4 Hollow Nanocubes Anchored on S-Doped Graphene for Ultrafast and Stable Na-Ion Storage.
SnS2 /Co3 S4 Hollow Nanocubes Anchored on S-Doped Graphene for Ultrafast and Stable Na-Ion Storage.
Small (Weinheim an der Bergstrasse, Germany) (2019-09-25)
Ya-Qian Wu, Hui-Xian Yang, Yu Yang, Hao Pu, Wen-Jie Meng, Rui-Ze Gao, Dong-Lin Zhao
ABSTRACT
SnS2 has been widely studied as an anode material for sodium-ion batteries (SIBs) based on the high theoretical capacity and layered structure. Unfortunately, rapid capacity decay associated with volume variation during cycling limits practical application. Herein, SnS2 /Co3 S4 hollow nanocubes anchored on S-doped graphene are synthesized for the first time via coprecipitation and hydrothermal methods. When applied as the anode for SIBs, the sample delivers a distinguished charge specific capacity of 1141.8 mAh g-1 and there is no significant capacity decay (0.1 A g-1 for 50 cycles). When the rate is increased to 0.5 A g-1 , it presents 845.7 mAh g-1 after cycling 100 times. Furthermore, the composite also exhibits an ultrafast sodium storage capability where 392.9 mAh g-1 can be obtained at 10 A g-1 and the charging time is less than 3 min. The outstanding electrochemical properties can be ascribed to the enhancement of conductivity for the addition of S-doped graphene and the existence of p-n junctions in the SnS2 /Co3 S4 heterostructure. Moreover, the presence of mesopores between nanosheets can alleviate volume expansion during cycling as well as being beneficial for the migration of Na+ .
Sigma-Aldrich
Tin(IV) chloride pentahydrate, 98%