Merck
  • Development of Membrane Hollow Fiber for Determination of Maleic Anhydride in Ambient Air as a Field Sampler.
Development of Membrane Hollow Fiber for Determination of Maleic Anhydride in Ambient Air as a Field Sampler.
Annals of work exposures and health (2019-07-07)
Ehsan Partovi, Abdulrahman Bahrami, Abbas AfKhami, Farshid Ghorbani Shahna, Farhad Ghamari, Maryam Farhadian
ABSTRACT
This research develops a rapid method for sampling and analysis of maleic anhydride (MA) in air using a one-step hollow fiber (HF) membrane in the liquid phase followed by high-performance liquid chromatography. A sampling chamber was prepared for sampling of MA with HF-supported de-ionized water absorbency. Several important parameters, such as sampling flow rate, sampling time, and breakthrough volume (BTV), were optimized at different concentrations using a central composite design. The results showed that sampling could be performed at the maximum period of 4 h with a flow rate of 1 mL min-1 for different concentrations (in the range of 0.05-2 mg m-3). The BTV was 240 mL. The relative standard deviations for the repeatability of interday and intraday were 7-10%, 10%, respectively, and the pooled standard deviation was 0.088. The limit of detection and limit of quantitation values were 0.033 and 0.060 mg m-3, respectively. Moreover, our findings revealed that the samples could be stored in sealed HF flexible plastic tubes in a cover at refrigerator temperature (4°C) for up to 7 days. The HF method was compared with method number 3512 National Institute Occupational Safety and Health for determination of MA. There was a good correlation (R2 = 0.99) between the two methods at a concentration of 0.05 to 2 mg m-3 in the laboratory and the average concentration of MA for both methods was 0.11 mg m-3 in the ambient air at an adhesive manufacturer. Our findings indicated that the proposed HF can act as a reliable, rapid, and effective approach for sampling of MA in workplaces.
Sigma-Aldrich
Dicyclohexylamine, 99%