Merck
  • Semaphorin 4C Promotes Macrophage Recruitment and Angiogenesis in Breast Cancer.
Semaphorin 4C Promotes Macrophage Recruitment and Angiogenesis in Breast Cancer.
Molecular cancer research : MCR (2019-07-17)
Jie Yang, Zhen Zeng, Long Qiao, Xuefeng Jiang, Jingjing Ma, Junnai Wang, Shuangmei Ye, Quanfu Ma, Juncheng Wei, Mingfu Wu, Xiaoyuan Huang, Ding Ma, Qinglei Gao
ABSTRACT
Semaphorins are a large family of evolutionarily conserved morphogenetic molecules that are associated with repelling axonal guidance. Intriguingly, recent researches indicate that semaphorins are involved in cancer progression. Semaphorin 4C (SEMA4C) has long been considered a neuronal migration gene, but we detected that it is also highly expressed in many malignant human cancers. During an investigation of subcutaneous tumor models, we found that SEMA4C expression promoted tumor growth and progression. We discovered that SEMA4C was involved in maintaining tumor cell self-renewal, likely by regulating the p53 pathway. Inhibiting the expression of endogenous SEMA4C in tumor cells impaired growth and induced senescence and cell-cycle arrest in the G2-phase. In addition, we found that SEMA4C induced the production of angiogenin and colony-stimulating factor-1 (CSF-1) in tumor cells by activating the NF-κB pathway in a plexinB2-dependent manner. In conclusion, SEMA4C expression in breast cancer cells promotes cancer cell proliferation, macrophage recruitment, and angiogenesis. Thus, inhibition of SEMA4C activity may be a novel therapeutic strategy for human breast cancer. IMPLICATIONS: In breast cancer, therapeutic targeting of the SEMA4C pathway may prevent tumor growth, angiogenesis, metastasis, and progression.
Sigma-Aldrich
In Vitro Vascular Permeability Assay (24-well), This In Vitro Vascular Permeability Assay kit employs a 24-well plate and provides an efficient system for evaluating the effects of chemicals & drug compounds on endothelial cell adsorption, transport & permeability.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)