Merck
Expression and structural analysis of 14-3-3 proteins.
Journal of molecular biology (1995-01-27)
D H Jones, H Martin, J Madrazo, K A Robinson, P Nielsen, P H Roseboom, Y Patel, S A Howell, A Aitken
ABSTRACT
The 14-3-3 family of proteins plays a role in a wide variety of cellular functions including regulation of protein kinase C and exocytosis. Using antisera specific for the N termini of 14-3-3 isoforms described previously and an additional antiserum specific for the C terminus of epsilon isoform, protease digestion of intact 14-3-3 showed that the N-terminal half of 14-3-3 (a 16 kDa fragment) was an intact, dimeric domain of the protein. Two isoforms of 14-3-3, tau and epsilon, were expressed in E. coli and their secondary structure was shown by circular dichroism to be identical to wild-type protein, and expression of N-terminally-deleted epsilon 14-3-3 protein showed that the N-terminal 26 amino acids are important for dimerization. Intact 14-3-3 is a potent inhibitor of protein kinase C, but the N-terminal domain does not inhibit PKC activity. Site-specific mutagenesis of several regions in the tau isoform of 14-3-3, including the mutation of a putative pseudosubstrate site to a potential substrate sequence, did not alter its inhibitory activity. Intact 14-3-3 proteins are phosphorylated by protein kinase C with a low stoichiometry, but truncated isoforms are phosphorylated much more efficiently by this kinase. This may imply that the proteins may adopt a different structural conformation, possibly upon binding to the membrane, which could modulate their activity. 14-3-3 proteins are found at high concentration on synaptic plasma membranes and this binding is mediated through the N-terminal 12 kDa of 14-3-3.