Merck
  • An RNA interference-based screen identifies MAP4K4/NIK as a negative regulator of PPARgamma, adipogenesis, and insulin-responsive hexose transport.
An RNA interference-based screen identifies MAP4K4/NIK as a negative regulator of PPARgamma, adipogenesis, and insulin-responsive hexose transport.
Proceedings of the National Academy of Sciences of the United States of America (2006-02-08)
Xiaoqing Tang, Adilson Guilherme, Abhijit Chakladar, Aimee M Powelka, Silvana Konda, Joseph V Virbasius, Sarah M C Nicoloro, Juerg Straubhaar, Michael P Czech
ABSTRACT
The insulin-regulated glucose transporter GLUT4 is a key modulator of whole body glucose homeostasis, and its selective loss in adipose tissue or skeletal muscle causes insulin resistance and diabetes. Here we report an RNA interference-based screen of protein kinases expressed in adipocytes and identify four negative regulators of insulin-responsive glucose transport: the protein kinases PCTAIRE-1 (PCTK1), PFTAIRE-1 (PFTK1), IkappaB kinase alpha, and MAP4K4/NIK. Integrin-linked protein kinase was identified as a positive regulator of this process. We characterized one of these hits, MAP4K4/NIK, and found that it is unique among mitogen-activated protein (MAP) kinases expressed in cultured adipocytes in attenuating hexose transport. Remarkably, MAP4K4/NIK suppresses expression of the adipogenic transcription factors C/EBPalpha, C/EBPbeta, and PPARgamma and of GLUT4 itself in these cells. RNA interference-mediated depletion of MAP4K4/NIK early in differentiation enhances adipogenesis and triglyceride deposition, and even in fully differentiated adipocytes its loss up-regulates GLUT4. Conversely, conditions that inhibit adipogenesis such as TNF-alpha treatment or depletion of PPARgamma markedly up-regulate MAP4K4/NIK expression in cultured adipocytes. Furthermore, TNF-alpha signaling to down-regulate GLUT4 is impaired in the absence of MAP4K4/NIK, indicating that MAP4K4 expression is required for optimal TNF-alpha action. These results reveal a MAP4K4/NIK-dependent signaling pathway that potently inhibits PPARgamma-responsive gene expression, adipogenesis, and insulin-stimulated glucose transport.
Sigma-Aldrich
HGK (1-328), active, GST tagged human, PRECISIOĀ® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ā‰„70% (SDS-PAGE), buffered aqueous glycerol solution