Skip to Content
Merck
SUPELCOSIL™ LC-CN HPLC Column 5 μm particle size, L × I.D. 15 cm × 4.6 mm

SUPELCOSIL LC-CN HPLC Column

SUPELCOSIL™ LC-CN HPLC Column 5 μm particle size, L × I.D. 15 cm × 4.6 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique