Phosphatidylcholine (Soy)
Synonym(s):
Phosphatidylcholine (Soy)
Empirical Formula (Hill Notation):
C42H80NO8P
CAS Number:
Molecular Weight:
758.06
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade