Cholecalciferol (Vitamin D3)
Synonym(s):
Cholecalciferol, (+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol, Vitamin D3 solution, 5,6-trans-vitamin D3, (3b,5E,7E)-9,10-Secocholesta-5,7,10(19)-trien-3-ol, calciol, cholecalciferol, colecalciferol
Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing