Skip to Content
Merck
Replacement 0.5 μm stainless steel frits for 2 mm I.D. TSKgel® columns pkg of 10 ea

Replacement 0.5 μm stainless steel frits for 2 mm I.D. TSKgel® columns

Replacement 0.5 μm stainless steel frits for 2 mm I.D. TSKgel® columns pkg of 10 ea
Synonym(s):
TSKgel® Hardware and Accessories

Products

Product Category
Manufacturer
Product Line
Pore size (µm)
Technique
Showing 1-1 of 1