Skip to Content
Merck
Poly(styrene-co-divinylbenzene) microspheres, 6.0-10.0 μm avg. part. size

Poly(styrene-co-divinylbenzene)

Poly(styrene-co-divinylbenzene) microspheres, 6.0-10.0 μm avg. part. size
Synonym(s):
Polystyrene crosslinked with divinylbenzene
Linear Formula:
[CH2CH(C6H5)]x[CH2CH[C6H4(CHCH2)]]y
CAS Number:

Products

Product Category
Brand
Physical Form
Separation Technique
Quality Segment
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing