Skip to Content
Merck
vibrant-m

Nizatidine for system suitability

vibrant-m
Synonym(s):
Nizatidine, N-{2-[2-(N,N-Dimethylaminomethyl)-4-thiazolylmethylthio]ethyl}-N′-methyl-2-nitro-1,1-ethenediamine
Empirical Formula (Hill Notation):
C12H21N5O2S2
CAS Number:
Molecular Weight:
331.46

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1