Monoclonal Anti-Dinitrophenyl antibody produced in mouse
Synonym(s):
Anti-Dinitrophenyl, Monoclonal Anti-DNP
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1