Hydrogen chloride solution
Synonym(s):
Hydrochloric acid in methanol, Hydrochloric acid in ethanol, Hydrogen chloride – ethanol solution, Hydrochloric acid in 2-propanol, HCl/Sulfuric Acid solution
Linear Formula:
HCl · CH3CH(OH)CH3
CAS Number:
Molecular Weight:
36.46
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Reaction Type
Reagent Type
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing