Skip to Content
Merck
Benzonase® Nuclease HC, Purity > 99% Effective Viscosity Reduction and Removal of Nucleic Acids from protein solutions

Benzonase® Nuclease HC, Purity > 99%

Benzonase® Nuclease HC, Purity > 99% Effective Viscosity Reduction and Removal of Nucleic Acids from protein solutions
Synonym(s):
Endonuclease from Serratia marcescens
CAS Number:

Products

Product Category
Brand
Biological Source
Physical Form
Recombinant Host
Quality Segment
Application