Skip to Content
Merck
vibrant-m

Ascentis® Express C8 Capillary, 2.7 μm HPLC Column

vibrant-m

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
Length (cm)
Quality Segment
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique