Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium bicarbonate
Synonym(s):
Sodium bicarbonate
Linear Formula:
NaHCO
3
CAS Number:
144-55-8
Molecular Weight:
84.01
EC Number:
205-633-8
Products
Product Category
salts (12)
buffers (4)
bases (1)
food additives (1)
Brand
SAFC (3)
Sigma-Aldrich (9)
Feature
Formula Weight
Manufacturer
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (2)
Anhydrous (1)
Food Grade (1)
Halal (1)
Kosher (1)
Puriss (1)
certified reference material (1)
pharmaceutical primary standard (1)
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
S5761
Sodium bicarbonate,
powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
S6014
Sodium bicarbonate,
ACS reagent, ≥99.7%
Expand
View Pricing
1.06329
Sodium bicarbonate,
for analysis EMSURE
®
ACS,Reag.Ph Eur
Expand
View Pricing
S8875
Sodium bicarbonate,
ReagentPlus
®
, ≥99.5%, powder
Expand
View Pricing
792519
Sodium bicarbonate,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99.7%
Expand
View Pricing
13433
Sodium bicarbonate,
puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Expand
View Pricing
1.37013
Sodium bicarbonate,
EMPROVE
®
EXPERT, Ph. Eur., BP, ChP, JP, USP
Expand
View Pricing
1.06323
Sodium bicarbonate,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, FCC, E 500
Expand
View Pricing
S6297
Sodium bicarbonate,
BioXtra, 99.5-100.5%
Expand
View Pricing
W700433
Sodium bicarbonate,
≥99%, FG
Expand
View Pricing
900506
Sodium bicarbonate,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥99.5%
Expand
View Pricing
1613655
Sodium bicarbonate,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
401676
Sodium bicarbonate,
−40-+140 mesh, ≥95%
Expand
View Pricing
1.72577
Sodium bicarbonate,
for table water, FCC, E 500
Expand
View Pricing
71631
Sodium bicarbonate,
tested according to Ph. Eur.
Expand
View Pricing
NIST2191A
Sodium bicarbonate,
NIST
®
SRM
®
2191a, pD Standard
Expand
View Pricing