Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Butanone
Synonym(s):
Ethyl methyl ketone, MEK, Methyl ethyl ketone
Linear Formula:
C
2
H
5
COCH
3
CAS Number:
78-93-3
Molecular Weight:
72.11
EC Number:
201-159-0
Products
Product Category
solvents (5)
Brand
Sigma-Aldrich (3)
Supelco (2)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (2)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
360473
2-Butanone,
ACS reagent, ≥99.0%
Expand
View Pricing
1.09708
2-Butanone,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
1.06014
2-Butanone,
(2-butanone) EMPLURA
®
Expand
View Pricing
676926
2-Butanone,
ACS reagent, ≥99.0%
Expand
View Pricing
34861-M
2-Butanone,
suitable for HPLC, ≥99.7%
Expand
View Pricing