Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium formate
Synonym(s):
Formic acid ammonium salt
Linear Formula:
HCO
2
NH
4
CAS Number:
540-69-2
Molecular Weight:
63.06
EC Number:
208-753-9
Products
Product Category
salts (2)
bases (1)
buffers (1)
Brand
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Anhydrous (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
70221
Ammonium formate,
eluent additive for LC-MS, LiChropur
™
, ≥99.0%
Expand
View Pricing
156264
Ammonium formate,
reagent grade, 97%
Expand
View Pricing
516961
Ammonium formate,
≥99.995% trace metals basis
Expand
View Pricing
09735
Ammonium formate,
BioUltra, ≥99.0% (calc. based on dry substance, NT)
Expand
View Pricing
798568
Ammonium formate,
anhydrous, free-flowing, Redi-Dri
™
, reagent grade, 97%
Expand
View Pricing