Sand
Synonym(s):
Sand, white quartz, Quartz, Silicon dioxide, Sea sand, QUARTZ FINE GRANULAR,, Silica, crystalline quartz
Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Length (mm)
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-20 of 24
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2