Formamide
Synonym(s):
Amide C1, Formic amide, Formamide, Formamide solution
Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Products
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing