Skip to Content
Merck
Cholecalciferol meets USP testing specifications

Cholecalciferol

Cholecalciferol meets USP testing specifications
Synonym(s):
(+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol, Vitamin D₃ (cholecalciferol) cryst. (1 g = 40 Mio.I.U.)
Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Technique
Showing 1-3 of 3