Skip to Content
Merck
vibrant-m

3,3′-Dichlorobenzidine solution

vibrant-m
Empirical Formula (Hill Notation):
C12H10Cl2N2
CAS Number:
Molecular Weight:
253.13

Products

Brand
Formula Weight
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique