Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Retinyl acetate
Synonym(s):
Retinol acetate, Vitamin A acetate
Empirical Formula (Hill Notation):
C
22
H
32
O
2
CAS Number:
127-47-9
Molecular Weight:
328.49
EC Number:
204-844-2
Products
Product Category
vitamins (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
R4632
Retinyl acetate,
synthetic, crystalline solid or supercooled liquid
Expand
View Pricing
R7882
Retinyl acetate,
solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Expand
View Pricing
R3250
Retinyl acetate,
synthetic, matrix dispersion, 475,000-650,000 USP units/g
Expand
View Pricing