Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Boron nitride
Linear Formula:
BN
CAS Number:
10043-11-5
Molecular Weight:
24.82
Products
Brand
Sigma-Aldrich (5)
Formula Weight
Physical Form
Purity
Quality Segment
Particle Size (µm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
255475
Boron nitride,
powder, ~1 μm, 98%
Expand
View Pricing
790532
Boron nitride,
nanopowder, <150 nm avg. part. size (TEM), 99% trace metals basis
Expand
View Pricing
912085
Boron nitride,
Multiwalled, powder, >90%
Expand
View Pricing
913332
Boron nitride,
Multiwalled, powder, >70%
Expand
View Pricing
919500
Boron nitride,
multiwalled, powder, >80%
Expand
View Pricing