Acetaminophen
Synonym(s):
4′-Hydroxyacetanilide, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, N-Acetyl-4-aminophenol, APAP, Paracetamol
Linear Formula:
CH3CONHC6H4(OH)
CAS Number:
Molecular Weight:
151.16
Products
Product Category
Brand
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing