Skip to Content
Merck
Calcium chloride dihydrate extra pure, for table water, FCC, E 509

Calcium chloride dihydrate

Calcium chloride dihydrate extra pure, for table water, FCC, E 509
Synonym(s):
Calcium chloride dihydrate
Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01

Products

Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique