Sodium phosphate monobasic monohydrate
Synonym(s):
Monosodium phosphate, Sodium dihydrogen phosphate monohydrate, Sodium phosphate monobasic monohydrate
Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
Products
Product Category
Brand
Color
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Showing 1-10 of 10
Compare
Product Number
Description
Pricing