Hyaluronic acid sodium salt from rooster comb
Synonym(s):
Poly(β-glucuronic acid-[1→3]-β-N-acetylglucosamine-[1→4]), alternating, Sodium hyaluronate, Hyaluronic Acid, Sodium Salt, Streptococcus pyrogenes
Linear Formula:
(C14H21NaNO11)n
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Quality Segment
Showing 1-19 of 19
Compare
Product Number
Description
Pricing