Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Petroleum benzine
Synonym(s):
Petroleum ether
CAS Number:
64742-49-0
Products
Product Category
solvents (11)
Brand
Sigma-Aldrich (1)
Supelco (10)
Physical Form
Product Line
Agency
Quality Segment
Special Grade
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
1.01775
Petroleum benzine,
for analysis boiling range 40-60°C EMSURE
®
ACS,ISO
Expand
View Pricing
32299-M
Petroleum benzine,
puriss. p.a., ACS reagent, reag. ISO, low boiling point hydrogen treated naphtha, bp≥ 90% 40-60 °C (≥ 90%)
Expand
View Pricing
1.01781
Petroleum benzine,
for analysis EMSURE
®
Reag. Ph Eur
Expand
View Pricing
1.01774
Petroleum benzine,
for analysis EMSURE
®
Expand
View Pricing
1.01772
Petroleum benzine,
for gas chromatography ECD and FID SupraSolv
®
Expand
View Pricing
1.01770
Petroleum benzine,
(naphtha benzine) EMPLURA
®
Expand
View Pricing
1.00910
Petroleum benzine,
EMPLURA
®
Expand
View Pricing
1.00915
Petroleum benzine,
boiling range to about 40°C EMPLURA
®
Expand
View Pricing
1.01777
Petroleum benzine,
for analysis EMSURE
®
Expand
View Pricing
1.01773
Petroleum benzine,
EMPLURA
®
Expand
View Pricing
1.01786
Petroleum benzine,
for analysis EMSURE
®
Expand
View Pricing