Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gentamicin sulfate salt
Synonym(s):
Gentamicin sulfate salt, Gentamycin Sulfate
CAS Number:
1405-41-0
Products
Product Category
antibiotics (3)
bioactive small molecules (1)
Brand
Sigma-Aldrich (4)
Biological Source
Color
Feature
Physical Form
Product Line
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
G1264
Gentamicin sulfate salt,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
G3632
Gentamicin sulfate salt,
potency: ≥590 I.U. Gentamicin base per mg
Expand
View Pricing
G1914
Gentamicin sulfate salt,
meets USP testing specifications, powder
Expand
View Pricing
345814-M
Gentamicin sulfate salt,
Broad-spectrum cell culture antibiotic that is nontoxic to viruses and mammalian cells at antibacterial and antimycoplasmal concentrations.
Expand
View Pricing