Skip to Content
Merck
All Photos(1)

Key Documents

SRP5106

Sigma-Aldrich

p18INK4C, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CDKN2C, INK4C, p18, p18-INK4C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~42 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CDKN2C(1031)

General description

p18INK4C is a member of the INK4 family of proteins that regulate the G1 to S cell cycle transition by binding to and inhibiting the pRb kinase activity of cyclin-dependent kinases 4 and 6. Mutation of p18INK4C impairs B-cell terminal differentiation and confers increased susceptibility to tumor development. p18INK4C can function as a tumor suppressor gene in Hodgkins lymphoma and its inactivation may contribute to the cell cycle deregulation and defective terminal differentiation characteristic of the Reed-Sternberg cells.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ravichandran N Venkataramani et al.
The Journal of biological chemistry, 277(50), 48827-48833 (2002-10-09)
p18(INK4c) is a member of the INK4 family of proteins that regulate the G(1) to S cell cycle transition by binding to and inhibiting the pRb kinase activity of cyclin-dependent kinases 4 and 6. The p16(INK4a) member of the INK4
Abel Sánchez-Aguilera et al.
Blood, 103(6), 2351-2357 (2003-12-03)
p18INK4c is a cyclin-dependent kinase (CDK) inhibitor that interferes with the Rb-kinase activity of CDK6/CDK4. Disruption of p18INK4c in mice impairs B-cell terminal differentiation and confers increased susceptibility to tumor development; however, alterations of p18INK4c in human tumors have rarely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service