Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9042-49

CY®5 Amidite 100mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000
NACRES:
NA.32

form

solid

manufacturer/tradename

Cytiva

color

Blue (Dark)

solubility

soluble (Soluble in cold water and hot water)

General description

CyDye fluors combine exceptional photostability with bright and intense signals, resulting in high sensitivity and a highly reproducible performance at a biologically suitable pH range of 3 to 10.

Amersham™ CyDye™ fluors are versatile fluorophores designed for a broad range of applications. The use of multicolor CyDye fluors enables multiplexing without crosstalk. In addition, the CyDye flours readily dissolve in aqueous buffers, which eliminates the need of organic solvents. The high purity and thorough QC testing of Amersham CyDye fluors generate high levels of chromophore and reactive dye content. Narrow excitation and emission bands result in discrete signals from each fluor, which together with minimal cross-talk contributes to high accuracy.

Features and Benefits

  • Wide selection of CyDye fluors for protein labeling for use in ELISA & immunoprecipitation.
  • Labeling strategy based on protein characteristics.
  • Medium to high-throughput fluorescence imaging.
  • Ready-to-use labeled antibodies enable multiplexed assays.
  • Streptavidin-conjugated CyDye fluors for use in Flow-cytometry and In situ hybridization.
  • Linkage of Cy dyes allows labeled DNA to hybridize to other DNA.
  • For incorporation of Cy5 fluorescent dyes at any position in an oligonucleotide during synthesis.
  • DNA synthesis reagent.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Other Notes

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials. Keep container tightly closed and sealed until ready for use

Legal Information

Cy is a registered trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service