G5250
β-D(+)Glucose
≥97% (remainder primarily α-anomer)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
≥97% (remainder primarily α-anomer)
impurities
≤3% α-anomer.
SMILES string
OCC1OC(O)C(O)C(O)C1O
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Kazuto Koike et al.
Biosensors, 10(6) (2020-06-04)
The characteristics of a glucose sensor based on an ion-sensitive TiO2/Ti extended gate electrode field-effect transistor (EGFET) are reported. A glucose oxidase-containing silk fibroin membrane was immobilized on a TiO2/Ti surface as the bio-sensing component. This EGFET-type biosensor was estimated
Cheng-Peng Sun et al.
Natural product research, 31(9), 1059-1065 (2016-12-31)
A new phenol glycoside, physanguloside A (1), was isolated from Physalis angulata together with four known compounds. We report herein, for the first time, the presence of compounds 2-5 in the genus Physalis. The structures of all the compounds were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service