Merck
  • Targeting osteomyelitis with complete [99mTc]besilesomab and fragmented [99mTc]sulesomab antibodies: kinetic evaluations.
Targeting osteomyelitis with complete [99mTc]besilesomab and fragmented [99mTc]sulesomab antibodies: kinetic evaluations.
The quarterly journal of nuclear medicine and molecular imaging : official publication of the Italian Association of Nuclear Medicine (AIMN) [and] the International Association of Radiopharmacology (IAR), [and] Section of the Society of... (2014-10-18)
Stefan Gratz, Patrik Reize, Bendix Kemke, Wim U Kampen, Markus Luster, Helmut Höffken
ZUSAMMENFASSUNG
The aim of this retrospective study was to compare the targeting of "pure" osteomyelitis (i.e., without surrounding soft tissue infection) by directly 99mTc-labelled complete immunoglobulin G (IgG) monoclonal antibody (MAb) ([99mTc]besilesomab) and by directly 99mTc-labelled fragment antigen-binding (FAb) MAb ([99mTc]sulesomab) in relation to their kinetic fate. A total of 73 patients with "pure" osteomyelitis were examined with [99mTc]besilesomab, (Scintimun®, IBA/CIS bio international, Saclay, France; N.=38) and [99mTc]sulesomab (LeukoScan®, Immunomedics Inc., Morris Plains, NJ, USA; N.=35). Kinetic data were deduced from whole-body and single-photon emission computed tomographic scans, performed 10 minutes to 24 hour p.i. (region-of-interest technique [ROI]). In targeting "pure" osteomyelitis, sensitivities at 1-4 hours were found to be higher for [99mTc]sulesomab (44% and 80% for [99mTc]besilesomab and [99mTc]sulesomab, respectively) but at significantly lower target/background (T/B) ratios than with [99mTc]besilesomab (1.8±0.3 versus 1.4±0.5 for [99mTc]besilesomab and [99mTc]sulesomab respectively; P<0.01). With [99mTc]besilesomab, there was a continuous osteomyelitis uptake over 24 hours, whereas with [99mTc]sulesomab, the maximal uptake occurred mostly within 1-4 hours, with subsequent clearance being slower for antigen-bound activity than for nonspecific background. Hence, diagnosis was possible mostly after 4h with [99mTc]sulesomab but often not before 24 hours with [99mTc]besilesomab, the later increasing significantly (P<0.01) in sensitivity (87% and 84% for [99mTc]besilesomab and [99mTc]sulesomab, respectively). These results show that the higher sensitivity of [99mTc]sulesomab in osteomyelitis targeting at earlier p.i. times does not rely on an increased antibody uptake but on a more rapid clearance of nonspecific background activity due to faster metabolism and excretion. Intact [99mTc]besilesomab show a slow, continuous uptake, leading to higher T/B at later p.i. times, often beyond the imaging possibilities of 99mTc.