Merck
  • Immunohistochemical and FISH analysis of MDM2 and CDK4 in a dedifferentiated extraskeletal osteosarcoma arising in the vastus lateralis muscle: differential diagnosis and diagnostic algorithm.
Immunohistochemical and FISH analysis of MDM2 and CDK4 in a dedifferentiated extraskeletal osteosarcoma arising in the vastus lateralis muscle: differential diagnosis and diagnostic algorithm.
Pathology, research and practice (2014-07-01)
Alexandra von Baer, Alexander Ehrhardt, Daniel Baumhoer, Regine Mayer-Steinacker, Markus Schultheiss, Thair Abdul-Nou, Thomas Mentzel, Falko Fend, Peter Möller, Gernot Jundt, Thomas F E Barth
ZUSAMMENFASSUNG
Extraskeletal osteosarcoma is a rare neoplasia within the broad differential diagnostic spectrum of calcifying intramuscular lesions. We present a case of a slowly increasing mass within the left vastus lateralis muscle. At first presentation the patient showed a partially calcified well defined mass with a diameter of 5 cm and with no direct contact to the femur. A biopsy from the periphery revealed an ossifying lesion compatible with myositis ossificans. The patient returned 18 months later with the lesion having increased to a diameter of 25 cm. The resection specimen revealed a well delimitated tumor with a central core of partially necrotic neoplastic bone. Besides, histology showed high mitotic areas with pleomorphic spindle cells and regions with cartilaginous differentiation. Immunohistochemistry demonstrated: vimentin+, CD34-, desmin-, actin-, EMA- and pancytokeratin- with focal S100 protein positivity and a Ki-67 index of 20%. Comparative genomic hybridization (CGH) revealed a gain of chromosomal material on 12q; FISH analyses for the CDK4 and MDM2 region showed high level amplifications. Consequently, a high-grade dedifferentiated extraskeletal osteosarcoma was diagnosed. In conclusion, analysis of the MDM2 and CDK4 status is a powerful and discriminating diagnostic tool to distinguish dedifferentiated extraskeletal osteosarcoma from other benign/malignant ossifying lesions in the skeletal muscle.