Merck
Primary gastric apoptosis-rich T-cell lymphoma co-expressing CD4, CD8, and cytotoxic molecules.
Virchows Archiv : an international journal of pathology (2000-06-02)
T F Barth, F Leithäuser, H Döhner, M Bentz, M Pawlita, U Schmid, P Möller
ZUSAMMENFASSUNG
In contrast to primary gastric lymphomas of B-cell type, little is known about primary gastric T-cell lymphomas. We describe three cases with remarkably similar features: diffuse growth, epitheliotropism, medium too large cell size, high apoptotic rates, and a CD3+, CD4+, CD8+, CD45RO+ immunophenotype. Clonal TCRgamma gene rearrangement was shown in two cases. Epstein-Barr virus infection was excluded in two cases. Taking advantage of fresh-frozen material, we analyzed two cases further, revealing CD5-, CD16+, CD56-, CD57-, CD25+, CD30+, CD103 (alphaEbeta7)+, bcl-2 protein+, CD95+, CD95 ligand(L)-. CD95L, however, was detected in histiocytic and fibroblastoid by stander cells. The lymphomas expressed granzyme B, perforin, and the TIA-1 antigen in various combinations. All three cases had a very unfavorable clinical course characterized by local recurrence and/or dissemination to other epithelial sites, leading to death within 6-12 months after the initial diagnosis despite surgery and aggressive antineoplastic treatment. These data suggest a novel variant of peripheral T-cell lymphoma operationally characterized as primary gastric, apoptosis-rich, CD103+, EBV-, T-cell lymphoma co-expressing CD4, CD8, CD16 and cytotoxic molecules.