Z708984
ART® non-filtered tip
HLT Ultramicro, size 0.5-20 μL, Pure packed, sterile
Synonym(e):
pipet tips, pipette tips
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Sterilität
sterile
Leistungsmerkmale
Pure packed
Verpackung
pack of 960 ea
package of 960 tips
Hersteller/Markenname
Molecular BioProducts 3521
Maximales Volumen
20 μL
Größe
0.5-20 μL
Verwandte Kategorien
Allgemeine Beschreibung
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Ultra-micro style is perfect for working with low-volume samples.
Rechtliche Hinweise
ART is a registered trademark of Molecular BioProducts, Inc.