Merck
Tissue and urinary KIM-1 relate to tumor characteristics in patients with clear renal cell carcinoma.
International urology and nephrology (2017-10-21)
Mirjana Mijuskovic, Ivan Stanojevic, Novak Milovic, Snezana Cerovic, Dejan Petrovic, Djoko Maksic, Bozidar Kovacevic, Tamara Andjelic, Predrag Aleksic, Brankica Terzic, Mirjana Djukic, Danilo Vojvodic
RESUMEN
The objective of this prospective follow-up trial was to ascertain whether the urinary kidney injury molecule-1 (uKIM-1) associates with tumor tissue (tKIM-1) expression and with the pathological characteristics of clear renal cell carcinoma (cRCC) in radically nephrectomized (RN) and/or in partially nephrectomized (PN) patients with cRCC, pre- and postoperatively. This clinical study included 40 patients subjected to RN/PN (cRCC group) and 30 healthy volunteers (control group). Urinary KIM-1 was determined by ELISA TIM-1/KIM-1 kit and normalized by urinary creatinine. Immunohistochemical staining (monoclonal anti-human anti-TIM-1/KIM-1/HAVCR antibody) was used for semiquantitative analysis of the tKIM-1 expression and expressed as a score (% KIM-1 positively stained tubules). Both markers were interpreted in terms of the tumor characteristics comprising tumor size, Fuhrman grade, pathological (pT) stage, tumor/nodes/metastasis (TNM) stage, lymphovascular invasion and type of surgery RN/PN. Preoperative uKIM-1 was significantly higher in the cRCC group compared to controls, such as uKIM-1 was statistically higher in RN than in PN patients. Postoperatively, uKIM-1 decreased to control values. Expression of tKIM-1 was documented in all nephrectomized patients. Significant associations were achieved between uKIM-1 and tKIM-1 and with considered tumor characteristics, especially with tumor size and grade. Based on the accomplished associations, we found uKIM-1 as a highly sensitive marker for cRCC diagnosis. The clinical trial registration number: 1110-2012.