Merck
Influence of Aluminium and EGCG on Fibrillation and Aggregation of Human Islet Amyloid Polypeptide.
Journal of diabetes research (2017-01-12)
Zhi-Xue Xu, Qiang Zhang, Gong-Li Ma, Cong-Heng Chen, Yan-Ming He, Li-Hui Xu, Yuan Zhang, Guang-Rong Zhou, Zhen-Hua Li, Hong-Jie Yang, Ping Zhou
RESUMEN
The abnormal fibrillation of human islet amyloid polypeptide (hIAPP) has been implicated in the development of type II diabetes. Aluminum is known to trigger the structural transformation of many amyloid proteins and induce the formation of toxic aggregate species. The (-)-epigallocatechin gallate (EGCG) is considered capable of binding both metal ions and amyloid proteins with inhibitory effect on the fibrillation of amyloid proteins. However, the effect of Al(III)/EGCG complex on hIAPP fibrillation is unclear. In the present work, we sought to view insight into the structures and properties of Al(III) and EGCG complex by using spectroscopic experiments and quantum chemical calculations and also investigated the influence of Al(III) and EGCG on hIAPP fibrillation and aggregation as well as their combined interference on this process. Our studies demonstrated that Al(III) could promote fibrillation and aggregation of hIAPP, while EGCG could inhibit the fibrillation of hIAPP and lead to the formation of hIAPP amorphous aggregates instead of the ordered fibrils. Furthermore, we proved that the Al(III)/EGCG complex in molar ratio of 1 : 1 as Al(EGCG)(H