Merck
Human CRB2 inhibits gamma-secretase cleavage of amyloid precursor protein by binding to the presenilin complex.
The Journal of biological chemistry (2010-03-20)
Yachiyo Mitsuishi, Hiroshi Hasegawa, Akinori Matsuo, Wataru Araki, Toshiharu Suzuki, Shinji Tagami, Masayasu Okochi, Masatoshi Takeda, Ronald Roepman, Masaki Nishimura
RESUMEN
Drosophila Crumbs has been reported to attenuate Notch signaling by inhibition of gamma-secretase cleavage at the wing margins. gamma-Secretase is an intramembrane protease that is responsible for the generation of amyloid-beta (Abeta) peptides from the beta-amyloid precursor protein (APP). Here, we re-examined gamma-secretase inhibition by human CRB2, which is the most abundant Crumbs ortholog in the brain. Transfected CRB2 inhibited proteolytic production of Abeta and APP intracellular domains from APP C-terminal fragments in HEK293 and SH-SY5Y cells. Conversely, knockdown of endogenous CRB2 increased gamma-secretase cleavage products in SH-SY5Y cells. CRB2 inhibition of gamma-cleavage was also detected in cell-free assays. CRB2 interacted with the gamma-secretase complex, but was not a competitive substrate for gamma-cleavage. The transmembrane domain of CRB2 was indispensable for inhibition of Abeta generation and mediated CRB2 binding with the gamma-secretase complex. In addition, the cytoplasmic domain appeared to play a supportive role in gamma-secretase inhibition, whereas mutational disruption of the two protein-binding motifs involved in the formation of cell adhesion complexes did not affect gamma-secretase inhibition. Co-overexpression of presenilin-1 or APH-1 abrogated gamma-secretase inhibition probably through prevention of the incorporation of CRB2 into the gamma-secretase complex. Our results suggest that CRB2 functions as an inhibitory binding protein that is involved in the formation of a mature but inactive pool of the gamma-secretase complex.