Merck
Quantitative detection of platelet GPIIb-IIIa receptor antagonist activity using a flow cytometric method.
Journal of clinical laboratory analysis (1998-07-22)
L J Green, P Marder, S L Um, J A Jakubowski, J B Lawrence
RESUMEN
Platelet-membrane surface receptors are important targets for pharmacologic intervention in cardiovascular disease. Among these, glycoprotein (GP) IIb-IIIa is dominant and integrally involved in platelet aggregation and thrombus formation. When activated, GPIIb-IIIa binds soluble fibrinogen (Fb) in a key, early step of this process. New drugs are under development that block Fb binding to GPIIb-IIIa and inhibit platelet aggregation. A thorough understanding of the relationship between circulating drug levels and the extent of GPIIb-IIIa receptor occupancy in humans is crucial for safe and efficacious use of these agents. Described here is the development of a new technique for measurement of GPIIb-IIIa receptor occupancy. In this assay, activated human platelets are incubated with biotinylated fibrinogen (Fb-biotin) followed by antibiotin-FITC.The extent of Fb binding is determined using flow cytometric analysis. Our results indicate that Fb-biotin binds rapidly to activated platelets and its detection is dependent on incubation temperature. Platelets that were pre-incubated with the GPIIb-IIIa antagonist echistatin were inhibited from binding Fb-biotin in a concentration-dependent manner. The fluorescence of processed samples was stable for two weeks in the cold. The assay described here is simple, cost effective, and can be adapted for use in clinical evaluations of these new drugs.