Merck
Physical training increases endothelial progenitor cells, inhibits neointima formation, and enhances angiogenesis.
Circulation (2003-12-24)
Ulrich Laufs, Nikos Werner, Andreas Link, Matthias Endres, Sven Wassmann, Kristina Jürgens, Eckart Miche, Michael Böhm, Georg Nickenig
RESUMEN
The molecular mechanisms by which physical training improves peripheral and coronary artery disease are poorly understood. Bone marrow-derived endothelial progenitor cells (EPCs) are thought to exert beneficial effects on atherosclerosis, angiogenesis, and vascular repair. To study the effect of physical activity on the bone marrow, EPCs were quantified by fluorescence-activated cell sorter analysis in mice randomized to running wheels (5.1+/-0.8 km/d, n=12 to 16 per group) or no running wheel. Numbers of EPCs circulating in the peripheral blood of trained mice were enhanced to 267+/-19%, 289+/-22%, and 280+/-25% of control levels after 7, 14, and 28 days, respectively, accompanied by a similar increase of EPCs in the bone marrow and EPCs expanded from spleen-derived mononuclear cells. eNOS-/- mice and wild-type mice treated with N(G)-nitro-l-arginine methyl ester showed lower EPC numbers at baseline and a significantly attenuated increase of EPC in response to physical activity. Exercise NO dependently increased serum levels of vascular endothelial growth factor and reduced the rate of apoptosis in spleen-derived EPCs. Running inhibited neointima formation after carotid artery injury by 22+/-2%. Neoangiogenesis, as assessed in a subcutaneous disc model, was increased by 41+/-16% compared with controls. In patients with stable coronary artery disease (n=19), moderate exercise training for 28 days led to a significant increase in circulating EPCs and reduced EPC apoptosis. Physical activity increases the production and circulating numbers of EPCs via a partially NO-dependent, antiapoptotic effect that could potentially underlie exercise-related beneficial effects on cardiovascular diseases.