Merck
  • Cloning of the RNA8 gene of Saccharomyces cerevisiae, detection of the RNA8 protein, and demonstration that it is essential for nuclear pre-mRNA splicing.
Cloning of the RNA8 gene of Saccharomyces cerevisiae, detection of the RNA8 protein, and demonstration that it is essential for nuclear pre-mRNA splicing.
Molecular and cellular biology (1988-03-01)
S P Jackson, M Lossky, J D Beggs
RESUMEN
Strains of Saccharomyces cerevisiae that bear the temperature-sensitive mutation rna8-1 are defective in nuclear pre-mRNA splicing at the restrictive temperature (36 degrees C), suggesting that the RNA8 gene encodes a component of the splicing machinery. The RNA8 gene was cloned by complementation of the temperature-sensitive growth defect of an rna8-1 mutant strain. Integrative transformation and gene disruption experiments confirmed the identity of the cloned DNA and demonstrated that the RNA8 gene encodes an essential function. The RNA8 gene was shown to be represented once per S. cerevisiae haploid genome and to encode a low-abundance transcript of approximately 7.4 kilobases. By using antisera raised against beta-galactosidase-RNA8 fusion proteins, the RNA8 gene product was identified in S. cerevisiae cell extracts as a low-abundance protein of approximately 260 kilodaltons. Immunodepletion of the RNA8 protein specifically abolished the activity of S. cerevisiae in vitro splicing extracts, confirming that RNA8 plays an essential role in splicing.