Merck
Phosphorylated p38, a negative prognostic biomarker, complements TNM staging prognostication in colorectal cancer.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-07-25)
Xin-Juan Fan, Xiang-Bo Wan, Xin-Hui Fu, Pei-Huang Wu, Dian-Ke Chen, Pu-Ning Wang, Li Jiang, Dao-Hai Wang, Zhi-Ting Chen, Yan Huang, Jian-Ping Wang, Lei Wang
RESUMEN
Phosphorylated p38 (p-p38) played a pivotal role in the regulation of disease progression and correlated with tumor prognosis. Here, we characterized the prognostic effect of p-p38 in colorectal cancer (CRC). Three hundred and sixteen CRC patients in stages I-III were recruited in this study. P-p38 expression was semi-quantitatively evaluated using tissue microarrays and immunohistochemistry staining. Overall survival (OS), disease-free survival (DFS), local failure-free survival (LFFS), and distant metastasis-free survival (DMFS) of patient subgroups, segregated by p-p38 expression level and clinical stage, were compared using Kaplan-Meier analysis. We found that p-p38 was overexpressed in 48.1 % (152/316) CRC tissues, whereas low or deficiently expressed in normal adjacent epithelia. Overexpression of p-p38 predicted poor OS (P < 0.001), DFS (P = 0.002), LFFS (P = 0.016), and DMFS (P = 0.025) in CRC. Importantly, patient subgroups in the early stage (stages I + II) and with low p-p38 had similar OS, PFS, LFFS, and DMFS probabilities to that of stage I, whereas those with high p-p38 were similar to stage III disease. In addition, for stage III disease, the subgroup with low p-p38 had a similar survival probability to that of stage I, whereas the subgroup with high p-p38 had the worst survival. Multivariate Cox analysis confirmed that p-p38 was indeed a significantly independent factor for death, recurrence, and distant metastases in CRC. Our results demonstrated that p-p38 was a negative independent prognostic factor for CRC. Complementing TNM staging with p-p38 might refine the risk definition more accurately for a subset of patients.