Merck
Maturation of mouse bone marrow dendritic cells (BMDCs) induced by Laminaria japonica polysaccharides (LJP).
International journal of biological macromolecules (2014-06-20)
Jingjuan Meng, Yan Cao, Yiming Meng, Hong Luo, Xinghua Gao, Fengping Shan
RESUMEN
The seaweed Laminaria japonica has been investigated in a laboratory research for its medical significance and LJP has been purified now. The objective of present study was to look at effect of LJP on structural, phenotypic and functional maturation of murine BMDCs. The structural maturation of BMDCs induced by LJP was evaluated by transmission electron microscopy (TEM); The phenotypic maturation of BMDCs was studied by flow cytometry(FCM) and functional maturation of BMDCs was analyzed by FITC-dextran, acid phosphatase (ACP) activity and enzyme linked immunosorbent assay (ELISA). We hereby proved that LJP markedly induced maturation of BMDCs with the data of decreased the number of lysosomes, upregulated expression of CD80, CD83, CD86, CD40 and MHC II key membrane molecules on BMDCs, downregulated phagocytosis, enriched production of IL-12 and TNF-α secreted by BMDCs. Therefore it should be concluded that LJP was with strong ability to induce maturation of BMDCs. Our data provided direct evidence to suggest that LJP could be considered as an immune stimulant in improving immune handicapped situation and as a useful adjuvant in vaccine designing.