Merck
  • Autologous bone marrow mononuclear cell therapy improves symptoms in patients with end-stage peripheral arterial disease and reduces inflammation-associated parameters.
Autologous bone marrow mononuclear cell therapy improves symptoms in patients with end-stage peripheral arterial disease and reduces inflammation-associated parameters.
Cytotherapy (2014-06-29)
Nasser M Malyar, Stefan Radtke, Khalil Malyar, Jawed Arjumand, Peter A Horn, Knut Kröger, Eva Freisinger, Holger Reinecke, Bernd Giebel, Franz-Eduard Brock
RESUMEN
The purpose of this study was to evaluate the effect of autologous bone marrow mononuclear cells (BM-MNCs) on symptoms and perfusion indices in severely symptomatic patients with peripheral arterial disease (PAD) without further option for endovascular or surgical revascularization. Only patients with severe symptomatic PAD (Fontaine class IIb-IV, Rutherford category 3-6) not amenable for revascularization were treated. Bone marrow from both cristae iliacae was harvested; MNCs were isolated by the Ficoll density-gradient method and transplanted by means of intra-arterial and intramuscular injection in the index limb. Functional (pain score, ulcer healing, maximum walking distance) and perfusion indices such as ankle-brachial-index and transcutaneous oxygen pressure were documented before and after BM-MNC therapy. Additionally, serum concentration of C-reactive protein and interleukin-6 were measured as markers of inflammation before and after BM-MNC treatment. Sixteen consecutive patients (four women; mean age, 63.0 ± 13 years) were treated with a mean dose of 4.2 ± 2.2 × 10(8) BM-MNCs. At 6 months' follow-up, ankle-brachial-index, transcutaneous oxygen pressure and maximum walking distance significantly increased, whereas C-reactive protein and interleukin-6 conversely decreased (P < 0.01 versus baseline values), resulting in 88% limb salvage, 75% pain reduction and 71% complete wound healing and/or reduction of ulcer size. One major and one minor amputation were performed, both in patients with Rutherford category 6. Autologous BM-MNC therapy in patients with end-stage PAD improves tissue perfusion indices and decreases markers of inflammation. If our observations could be confirmed by large-scale, randomized controlled trials, BM-MNC transplantation could become an alternative therapeutic option for patients with end-stage PAD.